Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Catalog number: J15694.AP
500 mL, Nalgene bottle, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Catalog number: J15694.AP
500 mL, Nalgene bottle, Each
Quantity
Packaging
Catalog number: J15694.AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6381-92-6
IUPAC Name
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Formula
C10H18N2Na2O10
InChI Key
OVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILES
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Protease activity
Passes test
DNase
(exo): Passes test
RNase
Passes test
DNase
(endo): Passes test
pH
8.0 ? 0.5

Description

Chelating agent used in many enzyme buffers and, at higher concentrations, as an enzyme inactivator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text