Thermo Scientific Chemicals

D-(+)-Galactose, >99%, Thermo Scientific Chemicals

Catalog number: J15995.A7
5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

D-(+)-Galactose, >99%, Thermo Scientific Chemicals

Catalog number: J15995.A7
5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: J15995.A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-23-4
IUPAC Name
6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular Formula
C6H12O6
InChI Key
WQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILES
OCC1OC(O)C(O)C(O)C1O
Residue on ignition
≤ 0.5%
Loss on Drying
≤ 0.5%
Form
White to off-white powder
Assay (HPLC)
≥ 99.0%
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text