Thermo Scientific Chemicals

Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals

Catalog number: J17886.06
5 g, Plastic bottle, Each
Thermo Scientific Chemicals

Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals

Catalog number: J17886.06
5 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: J17886.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
367-93-1
IUPAC Name
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
Molecular Formula
C9H18O5S
InChI Key
BPHPUYQFMNQIOC-NXRLNHOXSA-N
SMILES
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Optical Rotation
-33° to -29° (c=1 in water)
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Form
Powder or crystals or flakes
Assay from Suppliers CofA
≥99% (HPLC)

Description

  • Induces the lac operon in E. coli by binding and inactivating the lac repressor.
  • Selection for lac Y mutants.
  • Selection for recombinant plasmids containing β-galactosidase fusion proteins.
  • Induces the cellular content of lactose permease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Induces lac operon in E. coli by binding and inactivating lac repressor
  • Selection for lac Y mutants
  • Selection for recombinant plasmids containing beta-galactosidase fusion proteins
  • Induces cellular content of lactose permease
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text