Thermo Scientific Chemicals

Kanamycin Sulfate, Ultrapure, Thermo Scientific Chemicals

Catalog number: J17924.14
25 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Kanamycin Sulfate, Ultrapure, Thermo Scientific Chemicals

Catalog number: J17924.14
25 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: J17924.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25389-94-0
IUPAC Name
(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-3-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}oxane-3,4,5-triol; sulfuric acid
Molecular Formula
C18H38N4O15S
InChI Key
OOYGSFOGFJDDHP-KMCOLRRFSA-N
SMILES
OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Impurity content
Organic Impurities: Meets the requirements
Loss on Drying
≤ 4.0%
Form
White to off-white powder
Comment
Crystallinity: Meets the requirements
Comment
Identity (A,B,C): Passes tests

Description

A selectable marker for cosmid vectors used in transfection of mammalian cells.

Mode of Action: Binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selectable marker for cosmid vectors used in transfection of mammalian cells
  • Mode of Action: Binds to ribosomal components and inhibits protein synthesis
  • Kanamycin is inactivated by aminoglycoside phospho transferase (MW 25,000) in periplasmic space
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text