You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Buffers and Standards
Life Science Buffers
Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Element Search
Structure Search
Thermo Scientific Chemicals
Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog number:
J22638.AP
500 mL, Nalgene bottle, Each
Certificates
Thermo Scientific Chemicals
Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog number:
J22638.AP
500 mL, Nalgene bottle, Each
Quantity
1 L
100 mL
500 mL
Packaging
Nalgene bottle
Certificates
Catalog number:
J22638.AP
Price (USD)
Special offer:
56.65
66.00
Save
9.35
(14%)
Ends:
15-Oct-2024
Each
Quantity
-
Request bulk or custom quote
Add to cart
Save to list
Have Questions?
Contact Us
Chemical Identifiers
CAS
1185-53-1
IUPAC Name
hydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular Formula
C4H12ClNO3
InChI Key
QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(CO)(CO)CO
View More
Specifications
Specification Sheet
DNase
(endo): Passes test
pH
(25C): Passes test
RNase
Passes test
DNase
(exo): Passes test
Protease activity
Passes test
Description
RUO – Research Use Only
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search