Tris, ≥98%, Thermo Scientific Chemicals
Tris, ≥98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris, ≥98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic bottle
Catalog number J22675.A1
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
Add to cart
Tris, ≥98%, Thermo Scientific Chemicals
Catalog numberJ22675.A1
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point≥ 160?C
pH(5%, H₂O): 10.0 - 11.5
FormWhite to pale yellow crystalline powder
Assay (unspecified)≥ 98.0% (dry basis)
Identification (FTIR)Conforms
View more
RUO – Research Use Only