Not web orderable
The product J500: Plastic Jars is not orderable in your country at the moment. Please contact our Technical Support for possible alternatives.
Explore other products in this category
Storage Jars
Use storage jars for gathering and collecting samples, specimens, and lab supplies. For optimal clarity, plastic jars are optimal; their straight sides also enable full-access for complete sample removal. For withstanding wide temperature ranges, polypropylene plastic jars are optimal and usually autoclavable. These features make them excellent for cold-room use and refrigeration. For glass-like quality, polystyrene plastic jars offer chemical resistance to household oils and solvents, aqueous products, acids, alcohols, and alkalis. Products include Thermo Scientific™ Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closures, which are designed to protect against breakage and contamination while offering light weight, good clarity, and excellent chemical resistance.