Thermo Scientific Chemicals

Enrofloxacin, Thermo Scientific Chemicals

Catalog number: J60023.14
25 g, Each
Thermo Scientific Chemicals

Enrofloxacin, Thermo Scientific Chemicals

Catalog number: J60023.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60023.14
also known as J60023-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93106-60-6
IUPAC Name
1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Molecular Formula
C19H22FN3O3
InChI Key
SPFYMRJSYKOXGV-UHFFFAOYSA-N
SMILES
CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O
Purity
≥95.0%
Appearance (Color)
Pale yellow
Mass Spec
Conforms
Form
Powder

Description

Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.

Solubility
Soluble in chloroform. Slightly soluble in water. Also soluble in dilute KOH

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text