Thermo Scientific Chemicals

Methylguanidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J60033.14
25 g, Each
Thermo Scientific Chemicals

Methylguanidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J60033.14
25 g, Each
Quantity
Catalog number: J60033.14
also known as J60033-14
Price (USD)
Quantity
-
Have Questions?

Description

Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

Solubility
Soluble in water (50 mg/ml - clear, colorless to light yellow solution).

Notes
Hygroscopic, store away form water/moisture. Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text