Thermo Scientific Chemicals

Abamectin, Thermo Scientific Chemicals

Catalog number: J60039.06
5 g, Each
Thermo Scientific Chemicals

Abamectin, Thermo Scientific Chemicals

Catalog number: J60039.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60039.06
also known as J60039-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71751-41-2
IUPAC Name
(1'R,2S,4'S,5S,6R,8'R,10'E,12'S,13'S,14'E,16'E,20'R,21'R,24'S)-21',24'-dihydroxy-12'-{[(2R,4S,5S,6S)-5-{[(2S,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy}-4-methoxy-6-methyloxan-2-yl]oxy}-5,11',13',22'-tetramethyl-6-(propan-2-yl)-5,6-dihydro-3',7',19'-trioxaspiro[pyran-2,6'-tetracyclo[15.6.1.1⁴,⁸.0²⁰,²⁴]pentacosane]-10',14',16',22'-tetraen-2'-one; (1'R,2S,4'S,5S,6R,8'R,10'E,12'S,13'S,14'E,16'E,20'R,21'R,24'S)-6-(butan-2-yl)-21',24'-dihydroxy-12'-{[(2R,4S,5S,6S)-5-{[(2S,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy}-4-methoxy-6-methyloxan-2-yl]oxy}-5,11',13',22'-tetramethyl-5,6-dihydro-3',7',19'-trioxaspiro[pyran-2,6'-tetracyclo[15.6.1.1⁴,⁸.0²⁰,²⁴]pentacosane]-10',14',16',22'-tetraen-2'-one
Molecular Formula
C95H142O28
InChI Key
IBSREHMXUMOFBB-MVGRHBATSA-N
SMILES
CO[C@H]1C[C@H](O[C@H]2[C@H](C)O[C@H](C[C@@H]2OC)O[C@H]2[C@@H](C)C=CC=C3/CO[C@@H]4[C@H](O)C(C)=C[C@@H](C(=O)O[C@H]5C[C@@H](CC=C2/C)O[C@@]2(C5)O[C@H](C(C)C)[C@@H](C)C=C2)[C@]34O)O[C@@H](C)[C@@H]1O.CCC(C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](CC=C(C)[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)C=CC=C4/CO[C@@H]5[C@H](O)C(C)=C[C@@H](C(=O)O3)[C@]45O)O2)C=C[C@@H]1C
Assay (HPLC)
Avermectin B1: 75% min.
Assay (HPLC)
Avermectin B1a: 70% min.
Assay (unspecified)
0.2% max.
Loss on Drying
2.0% max.

Description

Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

Solubility
Soluble in DMSO

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text