Psoralen, 97%, Thermo Scientific Chemicals
Psoralen, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Psoralen, 97%, Thermo Scientific Chemicals

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure
Have Questions?
Quantity:
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60042.MC
also known as J60042-MC
Price (USD)/ Each
279.00
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
279.00
Add to cart
Psoralen, 97%, Thermo Scientific Chemicals
Catalog numberJ60042.MC
Price (USD)/ Each
279.00
-
Add to cart
Specifications
Chemical Name or MaterialPsoralen
CAS66-97-7
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only