L-Phenylalanyl-L-phenylalanine, Thermo Scientific Chemicals
L-Phenylalanyl-L-phenylalanine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Phenylalanyl-L-phenylalanine, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number J60043.03
also known as J60043-03
Price (USD)/ Each
151.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
151.00
Add to cart
L-Phenylalanyl-L-phenylalanine, Thermo Scientific Chemicals
Catalog numberJ60043.03
Price (USD)/ Each
151.00
-
Add to cart
Chemical Identifiers
CAS2577-40-4
IUPAC Name2-(2-amino-3-phenylpropanamido)-3-phenylpropanoic acid
Molecular FormulaC18H20N2O3
InChI KeyGKZIWHRNKRBEOH-UHFFFAOYNA-N
SMILESNC(CC1=CC=CC=C1)C(=O)NC(CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Carbon-13 NMRConforms to structure
Solution TestClear solution in AcOH (0.3 gm/ 2 ml)
Infrared spectrumConforms to structure
Optical Rotation|b = +41 ± 2? (C-1 in AcOH)
Assay (HPLC)≥ 89%
View more
Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes. These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes
  • These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation
RUO – Research Use Only