Melanocyte-Stimulating Hormone-Release Inhibiting Factor, synthetic
Melanocyte-Stimulating Hormone-Release Inhibiting Factor, synthetic
Thermo Scientific Chemicals
Melanocyte-Stimulating Hormone-Release Inhibiting Factor, synthetic
Inhibits α-melanocyte-stimulating hormone (α-MSH) release. Increases brain dopamine turnover and inhibits physiological and behavioral opioid effects in vivo. Role in vascular homeostasis.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250mg
1g
Catalog number J60045.MD
also known as J60045-MD
Price (USD)/ Each
60.70
-
Add to cart
Quantity:
250mg
Request bulk or custom format
Price (USD)/ Each
60.70
Add to cart
Melanocyte-Stimulating Hormone-Release Inhibiting Factor, synthetic
Catalog numberJ60045.MD
Price (USD)/ Each
60.70
-
Add to cart
Specifications
Molecular Weight (g/mol)284.36
Quantity250mg
PackagingGlass bottle
FormLyophilized
Unit SizeEach
Inhibits α-melanocyte-stimulating hormone (α-MSH) release. Increases brain dopamine turnover and inhibits physiological and behavioral opioid effects in vivo. Role in vascular homeostasis.