Thermo Scientific Chemicals

D-(+)-Glucose, 1M aq. soln., sterile-filtered, Thermo Scientific Chemicals

Catalog number: J60067.EQE
125 mL, Each
Thermo Scientific Chemicals

D-(+)-Glucose, 1M aq. soln., sterile-filtered, Thermo Scientific Chemicals

Catalog number: J60067.EQE
125 mL, Each
Quantity
Catalog number: J60067.EQE
also known as J60067-=E
Price (USD)
Quantity
-
Have Questions?

Description

D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.

Solubility
Not miscible or difficult to mix with water. Soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text