Hydroxylapatite, fast flow, Thermo Scientific Chemicals
Hydroxylapatite, fast flow, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydroxylapatite, fast flow, Thermo Scientific Chemicals

Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number J60076.18
also known as J60076-18
Price (USD)/ Each
335.65
Online exclusive
373.00 
Save 37.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
335.65
Online exclusive
373.00 
Save 37.35 (10%)
Add to cart
Hydroxylapatite, fast flow, Thermo Scientific Chemicals
Catalog numberJ60076.18
Price (USD)/ Each
335.65
Online exclusive
373.00 
Save 37.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1306-06-5
IUPAC Namepentacalcium hydroxide triphosphate
Molecular FormulaCa5HO13P3
InChI KeyXYJRXVWERLGGKC-UHFFFAOYSA-D
SMILES[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentDNA binding capacity: 700?g/g
CommentBSA binding capacity: 12mg/g min.
Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only