Thermo Scientific Chemicals

Alcian Blue 8GX

Catalog number: J60122.14
25 g, Each
Thermo Scientific Chemicals

Alcian Blue 8GX

Catalog number: J60122.14
25 g, Each
Quantity
Catalog number: J60122.14
also known as J60122-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33864-99-2
IUPAC Name
(9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride
Molecular Formula
C56H68Cl4CuN16S4
InChI Key
XRWQTDIELZDCPS-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C
Appearance (Color)
Dark blue to blue-green
Form
Powder or crystals or crystalline powder
Assay (Dye)
≥50%
lambda-max (UV-Vis)
610-635 nm
Solution Test
Clear blue solution (75 mg in 250 water)

Description

Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.

Solubility
Soluble in water, ethanol, ethylene glycol and 2-ethoxyethanol. Insoluble in xylene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text