Thermo Scientific Chemicals

2-Iminothiolane hydrochloride, Thermo Scientific Chemicals

Catalog number: J60131.MD
250 mg, Each
Thermo Scientific Chemicals

2-Iminothiolane hydrochloride, Thermo Scientific Chemicals

Catalog number: J60131.MD
250 mg, Each
Quantity
Catalog number: J60131.MD
also known as J60131-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4781-83-3
IUPAC Name
hydrogen thiolan-2-imine chloride
Molecular Formula
C4H8ClNS
InChI Key
ATGUDZODTABURZ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].N=C1CCCS1
Form
Powder
Solution Test
100mg/mL in water
Appearance (Color)
White
Assay (HPLC)
97.0% min.

Description

2-Iminothiolane hydrochloride is predominantly used as an amine reactive at pH 7-10 and is also an effective thiolation reagent for polysaccharides. It is also used as a RNA-protein crosslinking reagent as well as useful in cross-linking proteins..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iminothiolane hydrochloride is predominantly used as an amine reactive at pH 7-10 and is also an effective thiolation reagent for polysaccharides. It is also used as a RNA-protein crosslinking reagent as well as useful in cross-linking proteins..

Solubility
Freely soluble in water

Notes
Protect from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text