Thermo Scientific Chemicals

Everolimus, Thermo Scientific Chemicals

Catalog number: J60139.MA
10 mg, Each
Thermo Scientific Chemicals

Everolimus, Thermo Scientific Chemicals

Catalog number: J60139.MA
10 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60139.MA
also known as J60139-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
159351-69-6
IUPAC Name
(1R,9S,12S,15R,16Z,18R,19R,21R,23S,24E,26Z,28E,30S,32S,35R)-1,18-dihydroxy-12-[(2R)-1-[(1S,3R,4R)-4-(2-hydroxyethoxy)-3-methoxycyclohexyl]propan-2-yl]-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-11,36-dioxa-4-azatricyclo[30.3.1.0⁴,⁹]hexatriaconta-16,24,26,28-tetraene-2,3,10,14,20-pentone
Molecular Formula
C53H83NO14
InChI Key
HKVAMNSJSFKALM-CNPAPGRKSA-N
SMILES
CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)C=C(C)/[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)C=CC=C/C=C(C)/[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N3CCCC[C@H]3C(=O)O2)OC)CC[C@H]1OCCO
Form
Powder or crystals or crystalline powder or granules or amorphous solid
Appearance (Color)
White to off-white to pale yellow
Assay from Suppliers CofA
≥95.0%

Description

Everolimus is a rapamycin analog. It is used to treat certain types of kidney cancer, breast cancer and brain tumor. It prevents transplant rejection by decreasing the activity of the immune system. It also act as a kinase inhibitor. It is used in the treatment of subependymal giant cell astrocytoma (SEGA; a type of brain tumor) in adults and children 1 year of age and older who have TSC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethysulfoxide,ethanol and chloroform. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text