Thermo Scientific Chemicals

Uridine-5'-diphosphate trisodium salt, 98+%, Thermo Scientific Chemicals

Catalog number: J60141.MC
100 mg, Each
Thermo Scientific Chemicals

Uridine-5'-diphosphate trisodium salt, 98+%, Thermo Scientific Chemicals

Catalog number: J60141.MC
100 mg, Each
Quantity
Catalog number: J60141.MC
also known as J60141-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21931-53-3
IUPAC Name
trisodium ({[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Formula
C9H11N2Na3O12P2
InChI Key
GFASEGWJDXDCKZ-UHFFFAOYNA-K
SMILES
[Na+].[Na+].[Na+].OC1C(O)C(OC1COP([O-])(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O
Appearance (Color)
White to off-white
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥98.0% (HPLC)

Description

It has been used as a purinergic agonist to study its effects on absorptive cationic flux in cochlear outer sulcus cells (OSC) and vestibular transitional cells (VTC). Also been used for nucleoside diphosphatase (NDPase) activity assays in rabbit retinae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used as a purinergic agonist to study its effects on absorptive cationic flux in cochlear outer sulcus cells (OSC) and vestibular transitional cells (VTC). Also been used for nucleoside diphosphatase (NDPase) activity assays in rabbit retinae.

Solubility
Soluble in water to 100mM

Notes
Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text