Thermo Scientific Chemicals

Glycine, 0.2M buffer soln., pH 3.5

Catalog number: J60154.AP
500 mL, Each
Thermo Scientific Chemicals

Glycine, 0.2M buffer soln., pH 3.5

Catalog number: J60154.AP
500 mL, Each
Quantity
Catalog number: J60154.AP
also known as J60154-AP
Price (USD)
Quantity
-
Have Questions?

Description

Glycine, is used as a compound for polyacrylamide gel electrophoresis and Western blots. It is also used widely as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine; sc-296648) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. It is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine, is used as a compound for polyacrylamide gel electrophoresis and Western blots. It is also used widely as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine; sc-296648) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. It is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
It is soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text