TBE urea sample buffer (2X)
TBE urea sample buffer (2X)
Thermo Scientific Chemicals

TBE urea sample buffer (2X)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
10 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60186.AC
also known as J60186-AC
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
Add to cart
TBE urea sample buffer (2X)
Catalog numberJ60186.AC
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialSample Loading Buffer
ColorBlue
Concentration2X
Physical FormLiquid
Quantity25 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only