Thermo Scientific Chemicals

Loratadine, 98+%, Thermo Scientific Chemicals

Catalog number: J60190.06
5 g, Each
Thermo Scientific Chemicals

Loratadine, 98+%, Thermo Scientific Chemicals

Catalog number: J60190.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60190.06
also known as J60190-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79794-75-5
IUPAC Name
ethyl 4-{13-chloro-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}piperidine-1-carboxylate
Molecular Formula
C22H23ClN2O2
InChI Key
JCCNYMKQOSZNPW-UHFFFAOYSA-N
SMILES
CCOC(=O)N1CCC(CC1)=C1C2=CC=C(Cl)C=C2CCC2=CC=CN=C12
Comment
Heavy metals: 20ppm max.
Assay (HPLC)
98.0% min.
Loss on Drying
0.5% max.
Ash
20ppm max.

Description

Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is soluble in DMSO (50 mg/ml), ethanol (77 mg/ml at 25°C), water (<1mg/mL at 25°C), chloroform, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text