Sodium orthovanadate, 100mM aq. soln., Thermo Scientific Chemicals
Sodium orthovanadate, 100mM aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium orthovanadate, 100mM aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
Catalog number J60191.AE
also known as J60191-AE
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
Add to cart
Sodium orthovanadate, 100mM aq. soln., Thermo Scientific Chemicals
Catalog numberJ60191.AE
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialSodium orthovanadate
CAS13721-39-6
ColorColorless
Health Hazard 1H335
Recommended StorageKeep cold
View more
Sodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.

Solubility
It is soluble in water (50 mg/ml, )

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ai et al. Inhibition of small-intestinal sugar absorption mediated by sodium orthovanadate Na3VO4 in rats and its mechanisms. World J.Gastroenterol. 2004, 15,(24), 3612-5.
  2. Matsumoto et al. Sodium orthovanadate enhances proliferation of progenitor cells in the adult rat subventricular zone after focal cerebral ischemia. J.Pharmacol.Exp.Ther. 2006, 318,(3), 982-91.