L-Kynurenine, Thermo Scientific Chemicals
L-Kynurenine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Kynurenine, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mg
Catalog number J60199.MF
also known as J60199-MF
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
Add to cart
L-Kynurenine, Thermo Scientific Chemicals
Catalog numberJ60199.MF
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialL-Kynurenine
CAS2922-83-0
ColorYellow
Recommended StorageAmbient temperatures
RTECS NumberCY9049700
View more
Intermediate in the breakdown of tryptophan

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only