Thermo Scientific Chemicals

Theophylline, Thermo Scientific Chemicals

Catalog number: J60203.RAT
2.27 kg, Each
Thermo Scientific Chemicals

Theophylline, Thermo Scientific Chemicals

Catalog number: J60203.RAT
2.27 kg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60203.RAT
also known as J60203-R@
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-55-9
IUPAC Name
1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C7H8N4O2
InChI Key
ZFXYFBGIUFBOJW-UHFFFAOYSA-N
SMILES
CN1C2=C(NC=N2)C(=O)N(C)C1=O
Appearance (Color)
White to off-white
Form
Powder
Assay (HPLC)
97.0% min.
Loss on Drying
0.50% max.

Description

Theophylline has been used to study its effects on rat gene expression in the ubiquitin-proteasome pathway that regulates spermatogenesis and epididymal sperm quality. Theophylline has also been used as an internal standard for the measurement of plasma paracetamol levels in humans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Theophylline has been used to study its effects on rat gene expression in the ubiquitin-proteasome pathway that regulates spermatogenesis and epididymal sperm quality. Theophylline has also been used as an internal standard for the measurement of plasma paracetamol levels in humans.

Solubility
Soluble in ethanol (moderately), DMSO, alkali hydroxides, ammonia, dilute hyrochloric acid, nitric acid, dilute aqueous acid, and dilute aqueous base. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text