Calmodulin, Bovine Testes
This product is animal origin and can only be shipped within the United States
Calmodulin, Bovine Testes
Thermo Scientific Chemicals

Calmodulin, Bovine Testes

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 mg
1 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60231.EXC
also known as J60231-!C
Price (USD)/ Each
-
Quantity:
2.5 mg
Request bulk or custom format
Chemical Identifiers
CAS77107-46-1
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Solubility: 1mg/ml in water
Assay (unspecified)Amino acid analysis: 80% min.
Activates cyclic phosphodiesterase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C