L-Ornithine hydrochloride
L-Ornithine hydrochloride
Thermo Scientific Chemicals

L-Ornithine hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
250 g
Catalog number J60241.22
also known as J60241-22
Price (USD)/ Each
100.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
100.00
Add to cart
L-Ornithine hydrochloride
Catalog numberJ60241.22
Price (USD)/ Each
100.00
-
Add to cart
Chemical Identifiers
CAS3184-13-2
IUPAC Namehydrogen 2,5-diaminopentanoic acid chloride
Molecular FormulaC5H13ClN2O2
InChI KeyGGTYBZJRPHEQDG-UHFFFAOYNA-N
SMILES[H+].[Cl-].NCCCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder
Impurity contentSulfate: ≤0.02%
Optical Rotation+23 to +25°
Assay (special)Other Amino Acids: None Detected
Heavy Metals≤10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only