Mecamylamine hydrochloride, Thermo Scientific Chemicals
Mecamylamine hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mecamylamine hydrochloride, Thermo Scientific Chemicals

Nicotinyl acetylcholine receptor antagonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60257.MC
also known as J60257-MC
Price (USD)/ Each
944.00
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
944.00
Add to cart
Mecamylamine hydrochloride, Thermo Scientific Chemicals
Catalog numberJ60257.MC
Price (USD)/ Each
944.00
-
Add to cart
Specifications
Chemical Name or MaterialMecamylamine hydrochloride
CAS826-39-1
Health Hazard 1H301-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Nicotinyl acetylcholine receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only