Thermo Scientific Chemicals

Thalidomide, Thermo Scientific Chemicals

Catalog number: J60271.MC
100 mg, Each
Thermo Scientific Chemicals

Thalidomide, Thermo Scientific Chemicals

Catalog number: J60271.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60271.MC
also known as J60271-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-35-1
IUPAC Name
2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C13H10N2O4
InChI Key
UEJJHQNACJXSKW-UHFFFAOYNA-N
SMILES
O=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12
Assay (unspecified)
95.0% min.

Description

Teratogenic compound found to inhibit HIV-1 replication and FGF-induced angiogenesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text