Thermo Scientific Chemicals

L-Alanine, Cell Culture Reagent

Catalog number: J60279.36
500 g, Each
Thermo Scientific Chemicals

L-Alanine, Cell Culture Reagent

Catalog number: J60279.36
500 g, Each
Quantity
Catalog number: J60279.36
also known as J60279-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-41-7
IUPAC Name
(2S)-2-aminopropanoic acid
Molecular Formula
C3H7NO2
InChI Key
QNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILES
C[C@H](N)C(O)=O
Loss on Drying
≤0.5%
Appearance (Color)
White
Assay from Suppliers CofA
≥98.0%
Form
Powder or crystals or crystalline powder
Residue on ignition
≤0.2%

Description

It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

Solubility
Soluble in water 166.5 g/L..

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text