Nitrazine Yellow, Thermo Scientific Chemicals
Nitrazine Yellow, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitrazine Yellow, Thermo Scientific Chemicals

pH indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number J60281.06
also known as J60281-06
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
Add to cart
Nitrazine Yellow, Thermo Scientific Chemicals
Catalog numberJ60281.06
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
-
Add to cart
Chemical Identifiers
CAS5423-07-4
IUPAC Namedisodium 3-[(1E)-2-(2,4-dinitrophenyl)diazen-1-yl]-4-hydroxynaphthalene-2,7-disulfonate
Molecular FormulaC16H8N4Na2O11S2
InChI KeyXBLIFEQTVVSTIM-QIKYXUGXSA-L
SMILES[Na+].[Na+].OC1=C2C=CC(=CC2=CC(=C1N=NC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormPowder
Assay (unspecified)Dye content: 85% min.
Assay (unspecified)Visual transition interval: pH 6.0 yellow; pH 7.2 blue
Solution Test1mg/ml in water: clear, yellow-brown to brown solution
Nitrazine Yellow is used to test vaginal pH during late pregnancy to determine the breakage of the amniotic sac. To perform a fecal pH test for diagnosing intestinal infections or other digestive problems. In civil engineering, to determine the carbonatation spread in concrete structures and therefore assess the state of the rebar's passivation film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrazine Yellow is used to test vaginal pH during late pregnancy to determine the breakage of the amniotic sac. To perform a fecal pH test for diagnosing intestinal infections or other digestive problems. In civil engineering, to determine the carbonatation spread in concrete structures and therefore assess the state of the rebar′s passivation film.

Solubility
Dissolve in water at 1mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Angela M. Mills and D. B. Garrioch. Use Of The Nitrazine Yellow Swab Test In The Diagnosis Of Ruptured Membranes. BJOG: An International Journal of Obstetrics & Gynaecology. 1977, 84 (2), 138-140.
  2. Lien Van der Schueren; Karen Hemelsoet; Veronique Van Speybroeck; Karen De Clerck. The influence of a polyamide matrix on the halochromic behaviour of the pH-sensitive azo dye Nitrazine Yellow. Dyes and Pigments. 2012, 94 (3), 443-451.