Carbadox, 98+%, Thermo Scientific Chemicals
Carbadox, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carbadox, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60284.22
also known as J60284-22
Price (USD)/ Each
273.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
273.00
Add to cart
Carbadox, 98+%, Thermo Scientific Chemicals
Catalog numberJ60284.22
Price (USD)/ Each
273.00
-
Add to cart
Specifications
Chemical Name or MaterialCarbadox
CAS6804-07-5
Health Hazard 1H228-H302-H350
Health Hazard 2GHS H Statement
H228-H350-H302
Flammable solid.
May cause cancer.
Harmful if swallowed.
Health Hazard 3P201-P202-P210-P240-P241-P264b-P270-P281-P301+P312-P308+P313-P330-P370+P378q-P501c
View more
Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

Solubility
Soluble in 1N NaOH (50 mg/ml), and water (partly).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. T Hayashi, I Suenaga; N Narukawa; T Yamazaki. In vitro and in vivo activities of sedecamycin against Treponema hyodysenteriae. Antimicrobial Agents and Chemotherapy. 1988, 32 (4), 458-461.
  2. Torey Looft; Heather K Allen; Thomas A Casey; David P Alt; Thaddeus B Stanton. Carbadox has both temporary and lasting effects on the swine gut microbiota. Dyes and Pigments. 2014, 5, 276.