Thermo Scientific Chemicals

Mercaptoacetic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: J60290.14
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Mercaptoacetic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: J60290.14
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: J60290.14
also known as J60290-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
367-51-1
IUPAC Name
sodium 2-sulfanylacetate
Molecular Formula
C2H3NaO2S
InChI Key
GNBVPFITFYNRCN-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)CS
Form
Powder
Assay (Titration)
≥ 98%
Solution Test
Solubility: Clear solution (200 mg in 1 ml water)

Description

Reagent that protects tryptophan in amino acid analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text