Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, 0.5M aq. soln., pH 8.0, autoclaved

Catalog number: J60292.AK
250 mL, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, 0.5M aq. soln., pH 8.0, autoclaved

Catalog number: J60292.AK
250 mL, Each
Quantity
Catalog number: J60292.AK
also known as J60292-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-00-4
IUPAC Name
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular Formula
C10H16N2O8
InChI Key
KCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILES
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Comment
Sterility: autoclaved
Comment
Concentration: 0.5M
pH
8.0 ? 0.1

Description

Ethylenediaminetetraacetic acid is used in the treatment of lead poisoning, brain damage, cancer, rheumatoid arthritis, osteoarthritis, an eye condition called macular degeneration, diabetes, Alzheimer's disease, multiple sclerosis and Parkinson's disease. It acts as a chelating agent and used for the preparation of many solutions including DNA loading dye and resuspension buffer which is isolation of plasmid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, metals and copper.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text