Thermo Scientific Chemicals

Tween 20 washing buffer, 0.5% in PBS (10X)

Catalog number: J60304.AP
500 mL, Each
Thermo Scientific Chemicals

Tween 20 washing buffer, 0.5% in PBS (10X)

Catalog number: J60304.AP
500 mL, Each
Quantity
Catalog number: J60304.AP
also known as J60304-AP
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
Appearance (Color)
Clear colorless
pH
7.4 ± 0.15 (1X concentration)
Comment
All components are dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and autoclaved

Description

Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as Western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunologicals, and eventually for incubation solutions of immunoreagents (labeled antibodies) to reduce unspecific background.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text