Thermo Scientific Chemicals

Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade

Catalog number: J60316.09
10 g, Each
Thermo Scientific Chemicals

Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade

Catalog number: J60316.09
10 g, Each
Quantity
Catalog number: J60316.09
also known as J60316-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51805-45-9
IUPAC Name
3-[bis(2-carboxylatoethyl)phosphanyl]propanoate
Molecular Formula
C9H12O6P
InChI Key
PZBFGYYEXUXCOF-UHFFFAOYSA-K
SMILES
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
Assay from Supplier's CofA
≥98.0 %
Form
Crystalline powder or powder or crystals or solid
Appearance (Color)
White
Melting Point
176 to 186?C

Description

For reducing disulfide bonds in peptides and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text