Thermo Scientific Chemicals

Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 6.7, with alkaline buffer

Catalog number: J60331.AN
400 mL, Each
Thermo Scientific Chemicals

Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 6.7, with alkaline buffer

Catalog number: J60331.AN
400 mL, Each
Quantity
Catalog number: J60331.AN
also known as J60331-AN
Price (USD)
Have Questions?

Chemical Identifiers

CAS
136112-00-0
IUPAC Name
3-methylbutan-1-ol; phenol; trichloromethane
Molecular Formula
C12H19Cl3O2
InChI Key
ZYWFEOZQIUMEGL-UHFFFAOYSA-N
SMILES
ClC(Cl)Cl.CC(C)CCO.OC1=CC=CC=C1
pH
6.5-6.9 at 25°C
Appearance (Color)
Colorless
Form
Liquid (2-layer)
Assay (unspecified)
≥99.0%
DNase
None detected

Description

Phenol:Chloroform:Isoamyl alcohol 25:24:1 is used in the extraction of DNA for the generation of DNA minisatellite fingerprints. It is also used for purifying RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text