Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J60332.03
1 g, Each
Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J60332.03
1 g, Each
Quantity
Catalog number: J60332.03
also known as J60332-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207738-08-7
IUPAC Name
3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride
Molecular Formula
C16H22Cl2N2
InChI Key
NYNRGZULARUZCC-UHFFFAOYSA-N
SMILES
Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
Appearance (Color)
White
Form
Powder
Assay (unspecified)
≥97.5%
Ash
≤0.1%
Heavy Metals
≤5ppm

Description

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

Solubility
Soluble in DMSO (∼1.7 mg/ml), and 1:300 DMSO:PBS (pH 7.2) (0.03 mg/ml).

Notes
Moisture and light sensitive. Store away from oxidizing agents, heat, light, acids, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text