Thermo Scientific Chemicals

Adenosine-5'-triphosphate disodium salt hydrate, ultrapure, 98%

Catalog number: J60336.22
100 g, Each
Thermo Scientific Chemicals

Adenosine-5'-triphosphate disodium salt hydrate, ultrapure, 98%

Catalog number: J60336.22
100 g, Each
Quantity
Catalog number: J60336.22
also known as J60336-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34369-07-8
IUPAC Name
disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Formula
C10H14N5Na2O13P3
InChI Key
TTWYZDPBDWHJOR-WCYUCLFNNA-L
SMILES
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Appearance (Color)
White to pale yellow
Form
Powder or crystals or crystalline powder
Assay (unspecified)
≥97.5%
Water Content (Karl Fischer Titration)
10% max.

Description

Adenosine 5'-Triphosphate, is used as a p2 purinergicagonist. It is also used as a nucleotide, the 5'-triphosphate of deoxyadenosine activated precursor in DNA synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine 5′-Triphosphate, is used as a p2 purinergicagonist. It is also used as a nucleotide, the 5′-triphosphate of deoxyadenosine activated precursor in DNA synthesis.

Solubility
Soluble at 5% (w/v) in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away fom strong oxidizing agents. Stable under ordinary conditions. Light sensitive, Hygroscopic
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text