Thermo Scientific Chemicals

Astemizole, 99+%, Thermo Scientific Chemicals

Catalog number: J60339.MF
50 mg, Each
Thermo Scientific Chemicals

Astemizole, 99+%, Thermo Scientific Chemicals

Catalog number: J60339.MF
50 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60339.MF
also known as J60339-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68844-77-9
IUPAC Name
1-[(4-fluorophenyl)methyl]-N-{1-[2-(4-methoxyphenyl)ethyl]piperidin-4-yl}-1H-1,3-benzodiazol-2-amine
Molecular Formula
C28H31FN4O
InChI Key
GXDALQBWZGODGZ-UHFFFAOYSA-N
SMILES
COC1=CC=C(CCN2CCC(CC2)NC2=NC3=CC=CC=C3N2CC2=CC=C(F)C=C2)C=C1
Assay (HPLC)
>99.0%

Description

Astemizole, is used as a potent and selective H1-receptor antagonist that displays antihistamine and antimalarial activity. It is also been used as a chaperone to correct folding defects and restore protein function for some mutated forms of hERG channels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is soluble in DMSO (25 mg/ml ), ethanol (25 mg/ml), chloroform, methanol, and water (partly miscible).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storag
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text