Chondroitin sulfate, 90+%, Thermo Scientific Chemicals
This product is animal origin and can only be shipped within the United States
Chondroitin sulfate, 90+%, Thermo Scientific Chemicals
Actual product may vary
Thermo Scientific Chemicals

Chondroitin sulfate, 90+%, Thermo Scientific Chemicals

Form: Powder
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
J60341.03
also known as J60341-03
1 g
J60341.06
also known as J60341-06
5 g
J60341.14
also known as J60341-14
25 g
3 Options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60341.03
also known as J60341-03
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.50 
Save 4.85 (16%)
-
Add to cart
Quantity:
1 g
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.50 
Save 4.85 (16%)
Add to cart
Chondroitin sulfate, 90+%, Thermo Scientific Chemicals
Catalog numberJ60341.03
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.50 
Save 4.85 (16%)
-
Add to cart
Chondroitin sulfate is used in a wide variety of areas of research such as bioengineering. Chondroitin sulfate may be used as a biomaterial copolymer or surface derivatization reagent in the development of drug delivery vehicles, tissue engineering devices and bioscaffolds. Chondroitin sulfate may be used for the development of biocompatible structures such as hydrogels, sponges, biofilms, microspheres and micelles. Used in the treatment of coronary atherosclerosis, myocardial ischemia, myocardial infraction. Chondroitin sulfate is a sulfated (in different positions) glycosaminoglycan (GAG), composed of repeating and alternating disaccharide units of D-glucuronic acid and N-acetylgalactosamine. Depending on the position of sulfate, Chondroitin sulfate is named as chondroitin sulfate A (sulfate on carbon 4 of the N-acetylgalactosamine (GalNAc) sugar, C (carbon 6 of the GalNAc sugar), D (carbon 2 of the glucuronic acid and 6 of the GalNAc sugar) and E (carbons 4 and 6 of the GalNAc sugar). The CAS # given is the representation of core component of this product.
Solubility
Soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Wen-Li Gu; Pei-Hua Lu. [Chondroitin sulfate proteoglycans in neural development and regeneration]. Sheng li ke xue jin zhan [Progress in physiology].2007, 38 (2), 101-105.
  2. John G Kelton; Donald M Arnold; Shannon M Bates. Nonheparin anticoagulants for heparin-induced thrombocytopenia. New England Journal of Medicine. 2013, 368 (8), 737-744.
Specifications
Chemical Name or MaterialChondroitin sulfate
ColorWhite
Recommended StorageAmbient temperatures
EINECS Number232-696-9
TSCAYes
MDL NumberMFCD00146419
Quantity1 g
Merck Index14,2214
Synonymchondroitin sulfate, chondroitin sulphate, chonsurid, chondroitin sulfate c, chondroitin polysulfate, chondroitin sulfates, chondroitin 4-sulfate, chondroitin 6-sulfate, chondroitin, hydrogen sulfate, chondroitin sulfuric acid
Solubility InformationSoluble in water.
InChI KeyKXKPYJOVDUMHGS-OSRGNVMNSA-N
SMILESCC(=O)NC1C(C(C(OC1O)OS(=O)(=O)O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O
IUPAC Name(2S,3S,4S,5R,6R)-6-[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-sulfooxyoxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
PubChem CID24766
Percent Purity≥90%
Physical FormPowder
Unit SizeEach