Auramine O, 80% dye content
Auramine O, 80% dye content
Thermo Scientific Chemicals

Auramine O, 80% dye content

Fluorescent stain for acid-resistant bacteria in sputum
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
Catalog number J60343.18
also known as J60343-18
Price (USD)/ Each
81.50
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
81.50
Add to cart
Auramine O, 80% dye content
Catalog numberJ60343.18
Price (USD)/ Each
81.50
-
Add to cart
Chemical Identifiers
CAS2465-27-2
IUPAC Namehydrogen 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline chloride
Molecular FormulaC17H22ClN3
InChI KeyKSCQDDRPFHTIRL-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C1=CC=C(C=C1)C(=N)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Solubility: Soluble in alcohol & slightly soluble in water
FormYellow powder
Assay (Dye)Dye content: ≥ 80 %
Fluorescent stain for acid-resistant bacteria in sputum

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only