Bradykinin Acetate Salt
Bradykinin Acetate Salt
Thermo Scientific Chemicals

Bradykinin Acetate Salt

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J60345.MB
also known as J60345-MB
Price (USD)/ Each
218.00
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
218.00
Add to cart
Bradykinin Acetate Salt
Catalog numberJ60345.MB
Price (USD)/ Each
218.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Purification MethodHPLC
FormatPowder
Molecular Weight (g/mol)1120.21
Product TypeBradykinin acetate salt
Quantity25 mg
View more
Bradykinin Acetate elicits vasoconstriction, a systemic blood pressure reflex, a stimulation of respiration, and contraction of the nictitating membrane. Bradykinin plays an important role in the regulation of fluid and electrolyte balance, smooth muscle contraction, vasodilation and capillary permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C