Chloropyramine hydrochloride, Thermo Scientific Chemicals
Chloropyramine hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloropyramine hydrochloride, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60350.03
also known as J60350-03
Price (USD)/ Each
41.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
41.30
Add to cart
Chloropyramine hydrochloride, Thermo Scientific Chemicals
Catalog numberJ60350.03
Price (USD)/ Each
41.30
-
Add to cart
Specifications
Chemical Name or MaterialChloropyramine hydrochloride
CAS6170-42-9
ColorWhite
Recommended StorageAmbient temperatures
EINECS Number228-216-2
View more
Competitive reversible H-1 receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only