Thermo Scientific Chemicals

Sodium cacodylate, 0.1M buffer soln., pH 6.5, Thermo Scientific Chemicals

Catalog number: J60367.AD
50 mL, Each
Thermo Scientific Chemicals

Sodium cacodylate, 0.1M buffer soln., pH 6.5, Thermo Scientific Chemicals

Catalog number: J60367.AD
50 mL, Each
Quantity
Catalog number: J60367.AD
also known as J60367-AD
Price (USD)
Have Questions?

Description

Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

Solubility
Soluble in water (∼100 g/L) at 20°C, ethanol (400 mg/mL), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from strong oxidizing agents, strong bases.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text