Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals
Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals

A cryoprotectant, also useful in plant cell culture research
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
Catalog number J60382.14
also known as J60382-14
Price (USD)/ Each
24.65
Online exclusive
27.00 
Save 2.35 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
24.65
Online exclusive
27.00 
Save 2.35 (9%)
Add to cart
Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals
Catalog numberJ60382.14
Price (USD)/ Each
24.65
Online exclusive
27.00 
Save 2.35 (9%)
-
Add to cart
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
SpecificationsSpecification SheetSpecification Sheet
Assay (special)K-Value: 13 to 19
Identification (FTIR)Conforms to structure
Water Content (Karl Fischer Titration)≤ 5.0%
pH3.0 to 7.0 (5% solution in water)
Solution TestClear cream to light yellow (100 mg/ml solution in water)
View more
Polyvinylpyrrolidone is a polymer used as a pharmaceutical aid, complexing agent, and solubilizer. PVP was used as a plasma volume expander for trauma victims. It acts as diluent, solvent and cleaning agent in various kinds of industries. It also acts as an adjuvant, emulsifier, filtering aid, tableting aid, thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only