Thermo Scientific Chemicals

Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals

Catalog number: J60382.22
100 g, Each
Thermo Scientific Chemicals

Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals

Catalog number: J60382.22
100 g, Each
Quantity
Catalog number: J60382.22
also known as J60382-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-39-8
Molecular Formula
(C6H9NO)n
SMILES
*-CC(-*)N1CCCC1=O
MDL Number
MFCD01076626
Assay (special)
K-Value: 13 to 19
Identification (FTIR)
Conforms to structure
Water Content (Karl Fischer Titration)
≤ 5.0%
pH
3.0 to 7.0 (5% solution in water)
Solution Test
Clear cream to light yellow (100 mg/ml solution in water)

Description

Polyvinylpyrrolidone is a polymer used as a pharmaceutical aid, complexing agent, and solubilizer. PVP was used as a plasma volume expander for trauma victims. It acts as diluent, solvent and cleaning agent in various kinds of industries. It also acts as an adjuvant, emulsifier, filtering aid, tableting aid, thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text