Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate, 97%, Thermo Scientific Chemicals

Catalog number: J60387.MC
100 mg, Each
Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate, 97%, Thermo Scientific Chemicals

Catalog number: J60387.MC
100 mg, Each
Quantity
Catalog number: J60387.MC
also known as J60387-MC
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
White to pale cream
Form
Powder
Water Content (Karl Fischer Titration)
≤ 8.0%
Assay (special)
Spectrophotometric assay: ≥ 97.0 % (using enzyme reaction)
Assay (special)
Sodium: 8.0 - 12.0%

Description

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

Solubility
It is soluble in 10 mM NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C Light sensitive. Hygroscopic
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text