Thermo Scientific Chemicals

Formaldehyde, 4% in PBS

Catalog number: J60401.AP
500 mL, Each
Thermo Scientific Chemicals

Formaldehyde, 4% in PBS

Catalog number: J60401.AP
500 mL, Each
Quantity
Catalog number: J60401.AP
also known as J60401-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-00-0
IUPAC Name
formaldehyde
Molecular Formula
CH2O
InChI Key
WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES
C=O
Appearance (Color)
Clear colorless
Form
Liquid
Comment
Formaldehyde solution is made by dissolving it in 1X PBS, pH 7.4, followed by submicron filtration
pH
7.40 ± 0.20

Description

Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text