Epirubicin hydrochloride, Thermo Scientific Chemicals
Epirubicin hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Epirubicin hydrochloride, Thermo Scientific Chemicals

Cell-permeable anthracycline anti-tumor antibiotic
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60411.MA
also known as J60411-MA
Price (USD)/ Each
207.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
244.00 
Save 36.35 (15%)
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
207.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
244.00 
Save 36.35 (15%)
Add to cart
Epirubicin hydrochloride, Thermo Scientific Chemicals
Catalog numberJ60411.MA
Price (USD)/ Each
207.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
244.00 
Save 36.35 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialEpirubicin hydrochloride
CAS56390-09-1
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
Cell-permeable anthracycline anti-tumor antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only